Gene Transcripts

Gene Symbol
UBXN7
Entrez Gene ID
26043
Gene Name
UBX domain protein 7
Gene Aliases
UBXD7
Chromosome Location
Chr. 3: 196347659 - 196454323 on Build GRCh38
UniGene ID
Hs.518524
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015562.1 NP_056377.1 5 516
XM_011512670.2 XP_011510972.1 5 464
XM_011512671.2 XP_011510973.1 4 321
GenBank mRNA AB018337.1 - 5 444
AK026451.1 - 5 469
AK129880.1 - 5 469
AK303630.1 - 4 321
BC028986.1 - 4 348
BC047420.1 - 5 466
DA119173.1 - 5 516

View Full Product Details