Gene Transcripts

Gene Symbol
Magec2
Entrez Gene ID
317590
Gene Name
melanoma antigen family C, 2
Gene Aliases
Chromosome Location
Chr. X: 145354883 - 145362427 on Build Rnor_6.0
UniGene ID
Rn.8063
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025713.1 NP_001020884.1 4 1005
XM_008773568.1 XP_008771790.1 4 986
GenBank mRNA BC097464.1 - 4 1005

View Full Product Details