Gene Transcripts

Gene Symbol
Jph2
Entrez Gene ID
296345
Gene Name
junctophilin 2
Gene Aliases
Chromosome Location
Chr. 3: 159709998 - 159776534 on Build Rnor_6.0
UniGene ID
Rn.6206
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037974.1 NP_001033063.1 2 1106
XM_006235526.2 XP_006235588.1 2 1997
GenBank mRNA DQ304564.1 - 2 1106
FQ223967.1 - 2 1533

View Full Product Details