Gene Transcripts

Gene Symbol
KRTAP5-3
Entrez Gene ID
387266
Gene Name
keratin associated protein 5-3
Gene Aliases
KRTAP5-9, KRTAP5.3
Chromosome Location
Chr. 11: 1607565 - 1608463 on Build GRCh38
UniGene ID
Hs.532471
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012708.2 NP_001012726.1 1 815
GenBank mRNA AB126072.1 - 1 815
AJ628246.1 - 1 525

View Full Product Details