Gene Transcripts
- Gene Symbol
- RHOU
- Entrez Gene ID
- 58480
- Gene Name
- ras homolog family member U
- Gene Aliases
- ARHU, CDC42L1, G28K, WRCH1, hG28K
- Chromosome Location
- Chr. 1: 228644647 - 228746669 on Build GRCh38
- UniGene ID
- Hs.647774
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021205.5 |
NP_067028.1
|
2, 3 |
995 |
|
NR_037962.1 |
|
2, 3 |
453 |
|
GenBank mRNA |
AB051826.1 |
- |
2, 3 |
408 |
|
AB074878.1 |
- |
2, 3 |
468 |
|
AB209511.1 |
- |
3, 4 |
527 |
|
AF211836.1 |
- |
2, 3 |
951 |
|
AF251701.1 |
- |
2, 3 |
188 |
|
AF282258.1 |
- |
2, 3 |
568 |
|
AF378087.1 |
- |
2, 3 |
329 |
|
AK289971.1 |
- |
2, 3 |
581 |
|
BC040076.1 |
- |
2, 3 |
576 |
5768957 |
DA321957.1 |
- |
2, 3 |
454 |
|
View Full Product Details