Gene Transcripts

Gene Symbol
GRWD1
Entrez Gene ID
83743
Gene Name
glutamate rich WD repeat containing 1
Gene Aliases
CDW4, GRWD, RRB1, WDR28
Chromosome Location
Chr. 19: 48445773 - 48453907 on Build GRCh38
UniGene ID
Hs.400625
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031485.3 NP_113673.3 3 601
GenBank mRNA AF337808.1 - 3 380
AK074676.1 - 3 569
AK315625.1 - 3 393
AL713729.1 - 3 376
BC002440.2 - 3 376 3347366
BC036281.1 - 3 352
BC047351.1 - 3 352
EF011614.1 - 3 374

View Full Product Details