Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033678.3 | NP_001028850.2 | 7 | 829 | |
NM_001160389.1 | NP_001153861.1 | 7 | 835 | ||
NM_001160390.1 | NP_001153862.1 | 7 | 817 | ||
NM_001160393.1 | NP_001153865.1 | 6 | 627 | ||
NM_031472.3 | NP_113660.1 | 6 | 745 | ||
XM_005274345.4 | XP_005274402.1 | 7 | 752 | ||
XM_005274346.4 | XP_005274403.1 | 7 | 711 | ||
XM_005274347.4 | XP_005274404.1 | 6 | 628 | ||
XM_006718706.3 | XP_006718769.1 | 7 | 714 | ||
XM_006718707.3 | XP_006718770.1 | 7 | 661 | ||
XM_017018400.1 | XP_016873889.1 | 6 | 628 | ||
GenBank mRNA | AK026364.1 | - | 5 | 1539 | |
AY211494.1 | - | 7 | 785 | ||
BC005133.2 | - | 6 | 641 | 3836683 | |
BQ674339.1 | - | 6 | 567 |