Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033123.3 | NP_149114.2 | 4 | 420 | |
NR_073075.1 | 4 | 423 | |||
XM_005253519.2 | XP_005253576.1 | 4 | 413 | ||
XM_017020176.1 | XP_016875665.1 | 4 | 413 | ||
XM_017020177.1 | XP_016875666.1 | 4 | 413 | ||
XM_017020178.1 | XP_016875667.1 | 4 | 413 | ||
XM_017020179.1 | XP_016875668.1 | 4 | 408 | ||
XM_017020180.1 | XP_016875669.1 | 4 | 302 | ||
XM_017020181.1 | XP_016875670.1 | 4 | 367 | ||
GenBank mRNA | AF532185.1 | - | 4 | 345 | |
AK292279.1 | - | 4 | 418 | ||
AK314516.1 | - | 4 | 376 | ||
AY035866.1 | - | 4 | 382 | ||
DB038101.1 | - | 4 | 424 |