Gene Transcripts
- Gene Symbol
- NOSTRIN
- Entrez Gene ID
- 115677
- Gene Name
- nitric oxide synthase trafficking
- Gene Aliases
- DaIP2
- Chromosome Location
- Chr. 2: 168786539 - 168865339 on Build GRCh38
- UniGene ID
- Hs.189780
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039724.3 |
NP_001034813.2
|
6, 7 |
470 |
|
NM_001171631.1 |
NP_001165102.1
|
10, 11 |
1171 |
|
NM_001171632.1 |
NP_001165103.1
|
5, 6 |
386 |
|
NM_052946.3 |
NP_443178.2
|
5, 6 |
377 |
|
XM_006712233.3 |
XP_006712296.1
|
6, 7 |
421 |
|
XM_011510540.2 |
XP_011508842.1
|
6, 7 |
421 |
|
XM_017003278.1 |
XP_016858767.1
|
6, 7 |
421 |
|
XM_017003279.1 |
XP_016858768.1
|
5, 6 |
744 |
|
XM_017003280.1 |
XP_016858769.1
|
4, 5 |
236 |
|
GenBank mRNA |
AJ532842.1 |
- |
6, 7 |
413 |
|
AK002203.2 |
- |
6, 7 |
442 |
|
AK093444.1 |
- |
10, 11 |
1169 |
|
AK290254.1 |
- |
6, 7 |
436 |
|
BC014189.2 |
- |
5, 6 |
377 |
4650302 |
BC093072.1 |
- |
6, 7 |
417 |
30327154 |
BP334580.1 |
- |
6, 7 |
470 |
|
DQ402497.1 |
- |
5, 6 |
329 |
|
View Full Product Details