Gene Transcripts

Gene Symbol
VPS18
Entrez Gene ID
57617
Gene Name
VPS18, CORVET/HOPS core subunit
Gene Aliases
PEP3
Chromosome Location
Chr. 15: 40894410 - 40903975 on Build GRCh38
UniGene ID
Hs.23876
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020857.2 NP_065908.1 3 632
XM_011521843.2 XP_011520145.1 2 162
XM_011521844.2 XP_011520146.1 3 737
XM_017022447.1 XP_016877936.1 3 652
GenBank mRNA AB040908.1 - 3 607
AF308802.1 - 3 607
AL713725.1 - 3 632
BC001513.2 - 3 459 2822780

View Full Product Details