Gene Transcripts

Gene Symbol
GRIN3A
Entrez Gene ID
116443
Gene Name
glutamate ionotropic receptor NMDA type subunit 3A
Gene Aliases
GluN3A, NMDAR-L, NR3A
Chromosome Location
Chr. 9: 101569352 - 101738580 on Build GRCh38
UniGene ID
Hs.654783
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133445.2 NP_597702.2 9 7561
GenBank mRNA AB075853.1 - 9 7561
AL137422.1 - 1 772
BC167432.1 - 9 7561 8860109

View Full Product Details