Gene Transcripts

Gene Symbol
UBQLNL
Entrez Gene ID
143630
Gene Name
ubiquilin-like
Gene Aliases
Chromosome Location
Chr. 11: 5514393 - 5516726 on Build GRCh38
UniGene ID
Hs.10688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145053.4 NP_659490.4 1 1484
GenBank mRNA BC012183.1 - 1 1344
BC034977.2 - 1 1453 4822902

View Full Product Details