Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077503.1 | NP_001070971.1 | 2 | 167 | |
NM_001077504.1 | NP_001070972.1 | 2 | 167 | ||
NM_001077505.1 | NP_001070973.1 | 2 | 167 | ||
NM_001077506.1 | NP_001070974.1 | 2 | 167 | ||
NM_170685.2 | NP_733786.2 | 2 | 185 | ||
GenBank mRNA | AF515828.1 | - | 2 | 185 | |
AF515829.1 | - | 2 | 167 | ||
AF515830.1 | - | 2 | 167 | ||
AF515831.1 | - | 2 | 167 | ||
AF521560.1 | - | 2 | 185 | ||
AY471574.1 | - | 2 | 167 |