Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164637.2 | NP_001158109.1 | 9 | 1161 | |
NM_173627.4 | NP_775898.2 | 10 | 1296 | ||
XM_011524658.2 | XP_011522960.1 | 11 | 1562 | ||
XM_011524669.2 | XP_011522971.1 | 10 | 1379 | ||
XM_011524670.1 | XP_011522972.1 | 8 | 1255 | ||
XM_011524671.2 | XP_011522973.1 | 8 | 1222 | ||
XM_017024511.1 | XP_016880000.1 | 7 | 1039 | ||
XM_017024512.1 | XP_016880001.1 | 10 | 1474 | ||
XM_017024514.1 | XP_016880003.1 | 9 | 1144 | ||
XM_017024515.1 | XP_016880004.1 | 8 | 1104 | ||
XM_017024516.1 | XP_016880005.1 | 7 | 1007 | ||
XM_017024517.1 | XP_016880006.1 | 8 | 1272 | ||
XM_017024518.1 | XP_016880007.1 | 7 | 1020 | ||
XM_017024519.1 | XP_016880008.1 | 8 | 1255 | ||
XM_017024520.1 | XP_016880009.1 | 5 | 1306 | ||
GenBank mRNA | AK092539.1 | - | 9 | 1161 | |
AK096344.1 | - | 10 | 1339 | ||
AK096802.1 | - | 8 | 1072 | ||
AK123689.1 | - | 6 | 1023 | ||
BC037889.2 | - | 5 | 937 | ||
BC045824.1 | - | 5 | 937 | ||
BX647411.1 | - | 7 | 1067 |