Gene Transcripts

Gene Symbol
HIST4H4
Entrez Gene ID
121504
Gene Name
histone cluster 4, H4
Gene Aliases
H4/p
Chromosome Location
Chr. 12: 14770720 - 14771131 on Build GRCh38
UniGene ID
Hs.745457
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175054.2 NP_778224.1 1 140
GenBank mRNA AK311916.1 - 1 140
BC020884.1 - 1 140

View Full Product Details