Gene Transcripts

Gene Symbol
Cacna1a
Entrez Gene ID
25398
Gene Name
calcium channel, voltage-dependent, P/Q type, alpha 1A subunit
Gene Aliases
BccA1, Cav2.1, rbA-1
Chromosome Location
Chr. 19: 25453195 - 25749550 on Build Rnor_6.0
UniGene ID
Rn.87769
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012918.3 NP_037050.2 1 151
XM_008772361.1 XP_008770583.1 3 915
GenBank mRNA M64373.1 - 1 151

View Full Product Details