Gene Transcripts

Gene Symbol
Wwp1
Entrez Gene ID
107568
Gene Name
WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
8030445B08Rik, AIP5, SDRP1, Tiul1
Chromosome Location
Chr. 4: 19608296 - 19709004 on Build GRCm38
UniGene ID
Mm.474845
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276292.1 NP_001263221.1 24 6091
NM_177327.5 NP_796301.2 25 6253
GenBank mRNA AK049113.1 - 1 1483
BC051405.1 - 4 3570

View Full Product Details