Gene Transcripts

Gene Symbol
Garem
Entrez Gene ID
381126
Gene Name
GRB2 associated, regulator of MAPK1
Gene Aliases
C86169, Fam59a, Gm944, mKIAA4238
Chromosome Location
Chr. 18: 21127342 - 21300139 on Build GRCm38
UniGene ID
Mm.312276
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033445.2 NP_001028617.2 6 2936
GenBank mRNA AK148318.1 - 6 2937
AK162177.1 - 3 1061
AK220564.1 - 5 4231
BC049983.1 - 3 1914

View Full Product Details