Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001005265.1 | NP_001005265.1 | 2 | 487 | |
NM_001005330.1 | NP_001005330.1 | 487 | |||
NM_019301.2 | NP_062174.1 | 2 | 487 | ||
GenBank mRNA | BC061736.1 | - | 2 | 256 | |
D42114.1 | - | 2 | 296 | ||
D42115.1 | - | 2 | 232 | ||
L36532.1 | - | 2 | 255 |