Gene Transcripts

Gene Symbol
Madcam1
Entrez Gene ID
54266
Gene Name
mucosal vascular addressin cell adhesion molecule 1
Gene Aliases
Chromosome Location
Chr. 7: 12918294 - 12922424 on Build Rnor_6.0
UniGene ID
Rn.87175
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019317.1 NP_062190.1 3 519
XM_008765179.1 XP_008763401.1 3 831
GenBank mRNA D87840.1 - 3 519

View Full Product Details