Gene Transcripts

Gene Symbol
Cylc2
Entrez Gene ID
74914
Gene Name
cylicin, basic protein of sperm head cytoskeleton 2
Gene Aliases
4930488P18Rik
Chromosome Location
Chr. 4: 51216667 - 51229963 on Build GRCm38
UniGene ID
Mm.159368
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162865.1 NP_001156337.1 4 522
GenBank mRNA AK015651.1 - 4 523

View Full Product Details