Gene Transcripts

Gene Symbol
Cyb5b
Entrez Gene ID
80773
Gene Name
cytochrome b5 type B (outer mitochondrial membrane)
Gene Aliases
Cyb5m, omb5
Chromosome Location
Chr. 19: 39357814 - 39391680 on Build Rnor_6.0
UniGene ID
Rn.10249
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030586.2 NP_085075.1 2 265
GenBank mRNA BC072535.1 - 2 265
X96392.1 - 2 133
Y12517.1 - 2 310

View Full Product Details