Gene Transcripts

Gene Symbol
Pde11a
Entrez Gene ID
140928
Gene Name
phosphodiesterase 11A
Gene Aliases
Chromosome Location
Chr. 3: 62818502 - 63211843 on Build Rnor_6.0
UniGene ID
Rn.88630
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080893.1 NP_543169.1 1 712
GenBank mRNA AB059362.1 - 1 712

View Full Product Details