Gene Transcripts

Gene Symbol
Ust5r
Entrez Gene ID
171398
Gene Name
integral membrane transport protein UST5r
Gene Aliases
Chromosome Location
Chr. 1: 224457895 - 224533840 on Build Rnor_6.0
UniGene ID
Rn.81214
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134380.1 NP_599207.1 2 696
GenBank mRNA AJ132858.1 - 2 696

View Full Product Details