Gene Transcripts

Gene Symbol
Cdr2
Entrez Gene ID
12585
Gene Name
cerebellar degeneration-related 2
Gene Aliases
AA617262
Chromosome Location
Chr. 7: 120957036 - 120982312 on Build GRCm38
UniGene ID
Mm.1640
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007672.2 NP_031698.2 5 1549
GenBank mRNA AK171706.1 - 5 1504
AK215504.1 - 1 99
BC117530.1 - 5 1438 40106297
BC118511.1 - 5 1438 40106298
DT902919.1 - 1 96
U88588.1 - 6 1442

View Full Product Details