Gene Transcripts

Gene Symbol
Gadd45a
Entrez Gene ID
13197
Gene Name
growth arrest and DNA-damage-inducible 45 alpha
Gene Aliases
AA545191, Ddit1, GADD45
Chromosome Location
Chr. 6: 67035096 - 67037407 on Build GRCm38
UniGene ID
Mm.72235
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007836.1 NP_031862.1 4 579
GenBank mRNA AK144900.1 - 4 579
BC011141.1 - 4 559 4035898
L28177.1 - 4 579

View Full Product Details