Gene Transcripts

Gene Symbol
Cxcl1
Entrez Gene ID
14825
Gene Name
chemokine (C-X-C motif) ligand 1
Gene Aliases
Fsp, Gro1, KC, Mgsa, N51, Scyb1, gro
Chromosome Location
Chr. 5: 90891245 - 90893121 on Build GRCm38
UniGene ID
Mm.21013
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008176.3 NP_032202.1 4 646
GenBank mRNA AK140312.1 - 1 1571
BC037997.1 - 4 613
J04596.1 - 4 580

View Full Product Details