Gene Transcripts
- Gene Symbol
- Map1b
- Entrez Gene ID
- 17755
- Gene Name
- microtubule-associated protein 1B
- Gene Aliases
- A230055D22, AI843217, LC1, MAP5, Mtap-5, Mtap1b, Mtap5
- Chromosome Location
- Chr. 13: 99421464 - 99516602 on Build GRCm38
- UniGene ID
- Mm.4173
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008634.2 |
NP_032660.2
|
2, 3 |
409 |
|
GenBank mRNA |
AK157749.1 |
- |
1 |
181 |
|
AK158307.1 |
- |
2, 3 |
368 |
|
BC019183.1 |
- |
2, 3 |
341 |
|
BC037105.1 |
- |
2, 3 |
354 |
|
BC046287.1 |
- |
2, 3 |
330 |
|
BC052016.1 |
- |
2, 3 |
330 |
|
BC078457.1 |
- |
2, 3 |
335 |
|
BC138033.1 |
- |
2, 3 |
339 |
8861052 |
BC138034.1 |
- |
2, 3 |
339 |
8861053 |
X51396.1 |
- |
2, 3 |
347 |
|
View Full Product Details