Gene Transcripts

Gene Symbol
Psmc5
Entrez Gene ID
19184
Gene Name
protease (prosome, macropain) 26S subunit, ATPase 5
Gene Aliases
mSUG1
Chromosome Location
Chr. 11: 106256185 - 106263112 on Build GRCm38
UniGene ID
Mm.272361
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008950.1 NP_032976.1 4 247
GenBank mRNA AK010505.1 - 4 267
AK012294.1 - 4 279
AK135451.1 - 4 257
AK145703.1 - 4 251
BC030840.1 - 4 279 3994523
Z54219.1 - 4 247

View Full Product Details