Gene Transcripts

Gene Symbol
Zfp92
Entrez Gene ID
22754
Gene Name
zinc finger protein 92
Gene Aliases
Chromosome Location
Chr. X: 73410978 - 73429813 on Build GRCm38
UniGene ID
Mm.5060
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009566.4 NP_033592.2 6 2152
GenBank mRNA AJ627043.1 - 1 421
AK141522.1 - 6 2153
U47104.1 - 6 2172

View Full Product Details