Gene Transcripts
- Gene Symbol
- Papss1
- Entrez Gene ID
- 23971
- Gene Name
- 3'-phosphoadenosine 5'-phosphosulfate synthase 1
- Gene Aliases
- AI325286, Asapk, SK1
- Chromosome Location
- Chr. 3: 131564768 - 131643671 on Build GRCm38
- UniGene ID
- Mm.244912
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289477.1 |
NP_001276406.1
|
5, 6 |
704 |
|
NM_001289478.1 |
NP_001276407.1
|
5, 6 |
891 |
|
NM_001289479.1 |
NP_001276408.1
|
6, 7 |
1002 |
|
NM_011863.2 |
NP_035993.1
|
4, 5 |
714 |
|
GenBank mRNA |
AK146507.1 |
- |
4, 5 |
587 |
|
AK153296.1 |
- |
4, 5 |
584 |
|
AK160875.1 |
- |
6, 7 |
906 |
|
AK168982.1 |
- |
4, 5 |
587 |
|
AK169117.1 |
- |
5, 6 |
766 |
|
AK169124.1 |
- |
4, 5 |
581 |
|
AK169204.1 |
- |
5, 6 |
766 |
|
BC066055.1 |
- |
5, 6 |
770 |
6833756 |
U34883.1 |
- |
4, 5 |
591 |
|
View Full Product Details