Gene Transcripts

Gene Symbol
Foxo4
Entrez Gene ID
54601
Gene Name
forkhead box O4
Gene Aliases
Afxh, Mllt7, afx
Chromosome Location
Chr. X: 101254259 - 101260873 on Build GRCm38
UniGene ID
Mm.240299
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018789.2 NP_061259.1 2 1708
GenBank mRNA AB032770.1 - 2 1488
AB221664.1 - 2 1415
AF114260.1 - 2 1708
BC139275.1 - 2 1549 8862297
BC139304.1 - 2 1549 8862326

View Full Product Details