Gene Transcripts

Gene Symbol
Nfat5
Entrez Gene ID
54446
Gene Name
nuclear factor of activated T cells 5
Gene Aliases
AI225870, B130038B15Rik, CAG-8, CAG80, NFATL1, OREBP, TonEBP, mKIAA0827, nfatz
Chromosome Location
Chr. 8: 107293000 - 107379517 on Build GRCm38
UniGene ID
Mm.390057
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286260.1 NP_001273189.1 7, 8 1801
NM_018823.2 NP_061293.2 8, 9 1855
NM_133957.3 NP_598718.2 9, 10 1917
GenBank mRNA AF162853.1 - 7, 8 1712
AF200687.1 - 8, 9 1490
AF228706.1 - 5, 6 1555
AF369980.1 - 9, 10 1556
AF453571.2 - 9, 10 1525
AK134022.1 - 9, 10 1921

View Full Product Details