Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019546.5 | NP_062419.2 | 6, 7 | 856 | |
GenBank mRNA | AF222851.1 | - | 6, 7 | 843 | |
AK011020.1 | - | 6, 7 | 848 | ||
AK011039.1 | - | 6, 7 | 856 | ||
AK050141.1 | - | 6, 7 | 857 | ||
BC018182.1 | - | 6, 7 | 842 | 4161894 | |
U80019.1 | - | 6, 7 | 854 |