Gene Transcripts

Gene Symbol
Nit2
Entrez Gene ID
52633
Gene Name
nitrilase family, member 2
Gene Aliases
1190017B19Rik, D16Ertd502e
Chromosome Location
Chr. 16: 57156665 - 57167332 on Build GRCm38
UniGene ID
Mm.383203
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023175.1 NP_075664.1 3 275
GenBank mRNA AF284573.1 - 3 275
AK004533.1 - 3 259
AK004535.1 - 3 261
BC020153.1 - 3 264 3992930

View Full Product Details