Gene Transcripts

Gene Symbol
Tomm7
Entrez Gene ID
66169
Gene Name
translocase of outer mitochondrial membrane 7 homolog (yeast)
Gene Aliases
1110020J08Rik, AW047273, Tom7
Chromosome Location
Chr. 5: 23838944 - 23844145 on Build GRCm38
UniGene ID
Mm.291868
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025394.3 NP_079670.1 3 250
GenBank mRNA AK003864.1 - 3 213
AK008581.1 - 3 167
BC055352.1 - 3 228 5011269
BE647192.1 - 2 52
BU555283.1 - 3 253
BY773505.1 - 3 243

View Full Product Details