Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000402.4 | NP_000393.4 | 5 | 690 | |
NM_001042351.2 | NP_001035810.1 | 5 | 579 | ||
GenBank mRNA | AK292304.1 | - | 5 | 516 | |
AK302341.1 | - | 5 | 517 | ||
AL560686.3 | - | 5 | 564 | ||
BC000337.2 | - | 5 | 505 | 2822640 | |
M21248.1 | - | 4 | 466 | ||
X03674.1 | - | 5 | 922 |