Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168615.1 | NP_001162086.1 | 3 | 1003 | |
NM_145976.4 | NP_666088.1 | 3 | 939 | ||
GenBank mRNA | AK036643.1 | - | 3 | 856 | |
AK153654.1 | - | 3 | 735 | ||
AK170349.1 | - | 3 | 784 | ||
AK220206.1 | - | 1 | 490 | ||
BC005528.1 | - | 3 | 637 | 3495346 | |
BC027057.1 | - | 2 | 522 | 5029696 | |
BC031130.1 | - | 3 | 771 | 4954852 |