Product Details

SNP ID
rs76831595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:133825704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTACACTCAACTTTCCTGATGC[C/T]TGCAGAACAGGAGCTTTGTTGATAA
Phenotype
MIM: 615852 MIM: 616883
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB6B PubMed Links

Gene Details

Gene
RAB6B
Gene Name
RAB6B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016577.3 3767 UTR 3 NP_057661.3
XM_011512893.2 3767 UTR 3 XP_011511195.1
Gene
SRPRB
Gene Name
SRP receptor beta subunit
There are no transcripts associated with this gene.

View Full Product Details