Product Details

SNP ID
rs76196400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151089372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATAGCTGTGGTGGAAAATGCTGA[G/T]TTAATGGAGGAAGTGTGGGCTTCCA
Phenotype
MIM: 611318
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLRN1-AS1 PubMed Links

Gene Details

Gene
CLRN1-AS1
Gene Name
CLRN1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
MED12L
Gene Name
mediator complex subunit 12 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053002.5 Intron NP_443728.3
XM_006713487.3 Intron XP_006713550.1
XM_011512390.2 Intron XP_011510692.1
XM_011512394.2 Intron XP_011510696.1
XM_011512399.2 Intron XP_011510701.1
XM_017005676.1 Intron XP_016861165.1
XM_017005677.1 Intron XP_016861166.1
XM_017005678.1 Intron XP_016861167.1
XM_017005679.1 Intron XP_016861168.1
XM_017005680.1 Intron XP_016861169.1
XM_017005681.1 Intron XP_016861170.1

View Full Product Details