Product Details

SNP ID
rs77747486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43450498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTAATACTAGAACTATGCTGTA[C/T]TTTATGTGCACTATCTCACTTGATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM267 PubMed Links

Gene Details

Gene
TMEM267
Gene Name
transmembrane protein 267
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022483.4 Intron NP_071928.2
XM_005248333.4 Intron XP_005248390.1
XM_005248335.3 Intron XP_005248392.1
XM_006714486.3 Intron XP_006714549.1
XM_011514074.2 Intron XP_011512376.1
XM_011514075.2 Intron XP_011512377.1

View Full Product Details