Product Details

SNP ID
rs77984596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:82756284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCTTCCCCAATTATGAGTGAT[G/T]ATTATAGGCTTCTTCAAGTGCTAGC
Phenotype
MIM: 604918
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PCLO PubMed Links

Gene Details

Gene
PCLO
Gene Name
piccolo presynaptic cytomatrix protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014510.2 18009 Intron NP_055325.2
NM_033026.5 18009 UTR 3 NP_149015.2
XM_017012006.1 18009 UTR 3 XP_016867495.1
XM_017012007.1 18009 Intron XP_016867496.1

View Full Product Details