Product Details

SNP ID
rs75124808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:93101129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGGAACATATTTGCTACTTTTC[A/G]TATATCTCACTCCTTCCTTTTTCAC
Phenotype
MIM: 610456
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAMD9 PubMed Links

Gene Details

Gene
SAMD9
Gene Name
sterile alpha motif domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193307.1 5138 UTR 3 NP_001180236.1
NM_017654.3 5138 UTR 3 NP_060124.2

View Full Product Details