Product Details

SNP ID
rs76416666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:139026215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATCTGTGCAAAAACATGATTATG[C/T]ACGATGTGAACAAATATAGAAAGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZC3HAV1L PubMed Links

Gene Details

Gene
ZC3HAV1L
Gene Name
zinc finger CCCH-type containing, antiviral 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080660.3 1244 UTR 3 NP_542391.2
XM_006716176.3 1244 Intron XP_006716239.1
XM_011516688.2 1244 Intron XP_011514990.1
XM_011516689.2 1244 Intron XP_011514991.1

View Full Product Details