Product Details

SNP ID
rs76521101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:90625750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAGCCACTGGTATTTGGTTGA[C/T]TGCCTTTAACCAGAGAAGATTTCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM64 PubMed Links

Gene Details

Gene
TMEM64
Gene Name
transmembrane protein 64
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008495.3 1064 Missense Mutation AAT,AGT N355S NP_001008495.2
NM_001146273.1 1064 Missense Mutation AAT,AGT N303S NP_001139745.1

View Full Product Details