Product Details

SNP ID
rs76642901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:58416498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGATCTGATGGAAGAAGCTGAC[A/G]TTAGTGTGAACATCTTCAGGAATTC
Phenotype
MIM: 610686
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBXN2B PubMed Links

Gene Details

Gene
UBXN2B
Gene Name
UBX domain protein 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077619.1 Intron NP_001071087.1
XM_005251165.2 Intron XP_005251222.1
XM_005251166.1 Intron XP_005251223.1

View Full Product Details