Product Details

SNP ID
rs79434927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104569569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCTACCCACTGAGATACCCTGT[C/T]ATCATGAGCAAGGGTGCCTATGTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR13C8 PubMed Links

Gene Details

Gene
OR13C8
Gene Name
olfactory receptor family 13 subfamily C member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004483.1 402 Silent Mutation GTC,GTT V134V NP_001004483.1

View Full Product Details