Product Details
- SNP ID
-
rs78254742
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:12384336 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCAGTCACTTTCTGATCCATTATT[C/T]CCATTCTTTCAGCAACTATTTATTG
- Phenotype
-
MIM: 608120
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PARVA
PubMed Links
Gene Details
- Gene
- PARVA
- Gene Name
- parvin alpha
View Full Product Details