Product Details

SNP ID
rs76926886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:27497857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATCACAAAAGATGAAATCTGAA[C/T]CTGTTTTGCATTTATAATTTGATGT
Phenotype
MIM: 611468 MIM: 612332
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BDNF-AS PubMed Links

Gene Details

Gene
BDNF-AS
Gene Name
BDNF antisense RNA
There are no transcripts associated with this gene.

Gene
LIN7C
Gene Name
lin-7 homolog C, crumbs cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018362.3 1413 UTR 3 NP_060832.1

View Full Product Details