Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_173811.3 | 1213 | Intron | NP_776172.1 | ||
XM_011520025.1 | 1213 | Missense Mutation | AAA,AAT | K231N | XP_011518327.1 |
XM_011520026.2 | 1213 | Missense Mutation | AAA,AAT | K231N | XP_011518328.1 |
XM_011520027.1 | 1213 | Missense Mutation | AAA,AAT | K231N | XP_011518329.1 |
XM_011520028.2 | 1213 | Missense Mutation | AAA,AAT | K231N | XP_011518330.1 |
XM_011520029.2 | 1213 | Missense Mutation | AAA,AAT | K231N | XP_011518331.1 |
XM_017017602.1 | 1213 | Missense Mutation | AAA,AAT | K231N | XP_016873091.1 |
XM_017017603.1 | 1213 | Missense Mutation | AAA,AAT | K231N | XP_016873092.1 |
XM_017017604.1 | 1213 | Intron | XP_016873093.1 | ||
XM_017017605.1 | 1213 | Intron | XP_016873094.1 | ||
XM_017017606.1 | 1213 | Intron | XP_016873095.1 |